Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3R)-3-Aminobutan-1-ol 98%

Supplier: AMBEED, INC
Danger

A675589-10G A675589-1G A675589-25G A675589-5G
76980-974EA 9 USD InStock
76980-974 76980-976 76980-978 76980-980
(3R)-3-Aminobutan-1-ol 98%
(3R)-3-Aminobutan-1-ol
Precaución: for Research Use Only


Formula: C₄H₁₁NO
MDL Number: MFCD13184351
CAS Number: 61477-40-5
UN: 2734
ADR: 8,3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty