Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoquinoline-3-carboxamide 98%

Supplier: AMBEED, INC
Warning

A231552-1G A231552-250MG A231552-5G
76976-170EA 38 USD InStock
76976-170 76976-172 76976-174
Isoquinoline-3-carboxamide 98%
Isoquinoline-3-carboxamide
Precaución: for Research Use Only


Formula: C₁₀H₈N₂O
MDL Number: MFCD00160690
CAS Number: 50458-77-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty