Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-bromo-5-hydroxybenzoate 98%

Supplier: AMBEED, INC
Warning

A648428-100MG A648428-1G A648428-250MG A648428-25G A648428-5G
76980-790EA 5 USD InStock
76980-790 76980-792 76980-794 76980-796 76980-798
Methyl-3-bromo-5-hydroxybenzoate 98%
Methyl-3-bromo-5-hydroxybenzoate
Precaución: for Research Use Only


Formula: C₈H₇BrO₃
MW: 231.05
MDL Number: MFCD10566800
CAS Number: 192810-12-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty