Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pseudothiohydantoin 98%

Supplier: AMBEED, INC
Warning

A499696-10G A499696-1G A499696-25G A499696-5G
76979-854EA 11 USD InStock
76979-854 76979-856 76979-858 76979-860
Pseudothiohydantoin 98%
Pseudothiohydantoin
Precaución: for Research Use Only


Formula: C₃H₄N₂OS
MDL Number: MFCD00003186
CAS Number: 556-90-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty