Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1,5-dimethyl-1H-indazole 98%

Supplier: AMBEED, INC
Danger

A539121-100MG A539121-1G A539121-250MG A539121-5G
76980-162EA 6 USD InStock
76980-162 76980-164 76980-166 76980-168
4-Bromo-1,5-dimethyl-1H-indazole 98%
4-Bromo-1,5-dimethyl-1H-indazole
Precaución: for Research Use Only


Formula: C₉H₉BrN₂
MW: 225.09 g/mol
MDL Number: MFCD12028620
CAS Number: 1159511-77-9
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty