Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-6-hydroxypyrimidine 95%

Supplier: AMBEED, INC
Warning

A215406-100MG A215406-1G A215406-250MG A215406-25G A215406-5G
76975-660EA 11 USD InStock
76975-660 76975-662 76975-664 76975-666 76975-668
4-Bromo-6-hydroxypyrimidine 95%
4-Bromo-6-hydroxypyrimidine
Precaución: for Research Use Only


Formula: C₄H₃BrN₂O
MDL Number: MFCD11223276
CAS Number: 1086382-38-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty