Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 5-methyl-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 95%

Supplier: AMBEED, INC
Warning

A285000-100MG A285000-250MG
76977-496EA 92 USD InStock
76977-496 76977-498
tert-Butyl 5-methyl-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 95%
tert-Butyl 5-methyl-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate
Precaución: for Research Use Only


Formula: C₁₈H₂₄N₂O₃
MDL Number: MFCD20133911
CAS Number: 873779-31-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty