Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-nitroanisole 98%

Supplier: AMBEED, INC
Warning

A310799-100G A310799-10G A310799-1G A310799-25G A310799-500G A310799-5G A310799-1000G
76977-886EA 12 USD InStock
76977-886 76977-888 76977-890 76977-892 76977-894 76977-896 77672-582
5-Fluoro-2-nitroanisole 98%
5-Fluoro-2-nitroanisole
Precaución: for Research Use Only


Formula: C₇H₆FNO₃
MDL Number: MFCD00077541
CAS Number: 448-19-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty