Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibenzo[b,e]oxepin-11(6H)-one 97%

Supplier: AMBEED, INC
Warning

A160370-1G A160370-250MG A160370-25G A160370-5G
76973-560EA 10 USD InStock
76973-560 76973-562 76973-564 76973-566
Dibenzo[b,e]oxepin-11(6H)-one 97%
Dibenzo[b,e]oxepin-11(6H)-one
Precaución: for Research Use Only


Formula: C₁₄H₁₀O₂
MDL Number: MFCD00963963
CAS Number: 4504-87-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty