Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3,5-difluorobenzonitrile 95%

Supplier: AMBEED, INC
Warning

A247245-1G A247245-25G
76976-604EA 10 USD InStock
76976-604 76976-606
4-Chloro-3,5-difluorobenzonitrile 95%
4-Chloro-3,5-difluorobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₂ClF₂N
MDL Number: MFCD18415564
CAS Number: 144797-57-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty