Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-methyl-5-nitropyridine 97%

Supplier: AMBEED, INC
Warning

A167870-100G A167870-1G A167870-250MG A167870-25G A167870-5G
76973-864EA 524 USD InStock
76973-864 76973-866 76973-868 76973-870 76973-872
3-Bromo-4-methyl-5-nitropyridine 97%
3-Bromo-4-methyl-5-nitropyridine
Precaución: for Research Use Only


Formula: C6H5BrN2O2
MW: 217.0200 g/mol
MDL Number: MFCD11847767
CAS Number: 69872-15-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty