Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-chloro-4-iodopyridine 97%

Supplier: AMBEED, INC
Warning

A413807-100MG A413807-1G A413807-250MG A413807-5G
76979-190EA 14 USD InStock
76979-190 76979-192 76979-194 77673-008
2-Bromo-5-chloro-4-iodopyridine 97%
2-Bromo-5-chloro-4-iodopyridine
Precaución: for Research Use Only


Formula: C₅H₂BrClIN
MDL Number: MFCD16295248
CAS Number: 1061357-88-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty