Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Phenyl-1,2,4-oxadiazol-5-ol 95%

Supplier: AMBEED, INC
Warning

A208824-1G A208824-250MG A208824-25G A208824-5G
76975-462EA 36 USD InStock
76975-462 76975-464 76975-466 76975-468
3-Phenyl-1,2,4-oxadiazol-5-ol 95%
3-Phenyl-1,2,4-oxadiazol-5-ol
Precaución: for Research Use Only


Formula: C₈H₆N₂O₂
MDL Number: MFCD17676121
CAS Number: 1456-22-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty