Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,4S)-Methyl 4-((tert-butoxycarbonyl)amino)pyrrolidine-2-carboxylate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A223375-100MG A223375-250MG
76975-898EA 93 USD InStock
76975-898 76975-900
(2R,4S)-Methyl 4-((tert-butoxycarbonyl)amino)pyrrolidine-2-carboxylate hydrochloride 95%
(2R,4S)-Methyl 4-((tert-butoxycarbonyl)amino)pyrrolidine-2-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₂₁ClN₂O₄
MDL Number: MFCD11042659
CAS Number: 1279032-67-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty