Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Dimethylamino)piperidine dihydrochloride 97%

Supplier: AMBEED, INC
Warning

A125383-100G A125383-10G A125383-1G A125383-5G
76971-836EA 143 USD InStock
76971-836 76971-838 76971-840 76971-842
4-(Dimethylamino)piperidine dihydrochloride 97%
4-(Dimethylamino)piperidine dihydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₈Cl₂N₂
MDL Number: MFCD00035296
CAS Number: 4876-59-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty