Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-Epicatechin 97%

Supplier: AMBEED, INC
Warning

A121952-1G A121952-250MG A121952-25G A121952-5G
76971-574EA 6.37 USD InStock
76971-574 76971-576 76971-578 76971-580 76971-582
(-)-Epicatechin 97%
(-)-Epicatechin
Precaución: for Research Use Only


Formula: C₁₅H₁₄O₆
MDL Number: MFCD00075648
CAS Number: 490-46-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty