Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-[2-(Methylsulfanyl)-4-pyrimidinyl]-1-ethanone 95%

Supplier: AMBEED, INC
Warning

A264495-100MG A264495-1G A264495-250MG A264495-5G
76977-038EA 8 USD InStock
76977-038 76977-040 76977-042 76977-044
1-[2-(Methylsulfanyl)-4-pyrimidinyl]-1-ethanone 95%
1-[2-(Methylsulfanyl)-4-pyrimidinyl]-1-ethanone
Precaución: for Research Use Only


Formula: C₇H₈N₂OS
MW: 168.22 g/mol
MDL Number: MFCD09263472
CAS Number: 496863-48-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty