Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Leucic acid 98%

Supplier: AMBEED, INC
Warning

A635111-100G A635111-25G
76980-700EA 250 USD InStock
76980-700 77673-724
DL-Leucic acid 98%
DL-Leucic acid
Precaución: for Research Use Only


Formula: C₆H₁₂O₃
MW: 132.16 g/mol
MDL Number: MFCD00004246
CAS Number: 498-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty