Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-methoxyphenylacetonitrile 95%

Supplier: AMBEED, INC
Warning

A260255-100MG A260255-1G A260255-250MG A260255-5G
76976-914EA 27 USD InStock
76976-914 76976-916 76976-918 76976-920
2-Fluoro-6-methoxyphenylacetonitrile 95%
2-Fluoro-6-methoxyphenylacetonitrile
Precaución: for Research Use Only


Formula: C₉H₈FNO
MDL Number: MFCD00671772
CAS Number: 500912-18-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty