Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Foretinib 98%

Supplier: AMBEED, INC
Warning

A169062-1MG A169062-5MG
76973-904EA 9 USD InStock
76973-904 76973-906
Foretinib 98%
Foretinib
Precaución: for Research Use Only


Formula: C₃₄H₃₄F₂N₄O₆
MDL Number: MFCD16038048
CAS Number: 849217-64-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty