Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thieno[2,3-b]pyridine 97%

Supplier: AMBEED, INC
Warning

A148552-1G A148552-250MG A148552-5G
76972-990EA 48 USD InStock
76972-990 76972-992 76972-994
Thieno[2,3-b]pyridine 97%
Thieno[2,3-b]pyridine
Precaución: for Research Use Only


Formula: C₇H₅NS
MDL Number: MFCD01631145
CAS Number: 272-23-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty