Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-chloroanisole 98%

Supplier: AMBEED, INC
Warning

A395824-100G A395824-10G A395824-5G A395824-25G
76978-994EA 180 USD InStock
76978-994 76978-996 76978-998 77672-958
4-Bromo-3-chloroanisole 98%
4-Bromo-3-chloroanisole
Precaución: for Research Use Only


Formula: C₇H₆BrClO
MW: 221.48
MDL Number: MFCD00070742
CAS Number: 50638-46-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty