Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ki 16425 98+%

Supplier: AMBEED, INC
Warning

A114375-10MG A114375-1MG A114375-50MG A114375-5MG
76971-092EA 36 USD InStock
76971-092 76971-094 76971-096 76971-098
Ki 16425 98+%
Ki 16425
Precaución: for Research Use Only


Formula: C₂₃H₂₃ClN₂O₅S
MW: 474.96 g/mol
MDL Number: MFCD06798341
CAS Number: 355025-24-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty