Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TRAM 34 98%

Supplier: AMBEED, INC
Warning

A338045-100MG A338045-250MG A338045-50MG
76978-364EA 166 USD InStock
76978-364 76978-366 76978-368
TRAM 34 98%
TRAM 34
Precaución: for Research Use Only


Formula: C₂₂H₁₇ClN₂
MDL Number: MFCD09842562
CAS Number: 289905-88-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty