Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-(+)-Glucose 98%

Supplier: AMBEED, INC
Warning

A185706-1000G A185706-100G A185706-25G A185706-500G
76974-602EA 15 USD InStock
76974-602 76974-604 76974-606 76974-608
D-(+)-Glucose 98%
D-(+)-Glucose
Precaución: for Research Use Only


Formula: C₆H₁₂O₆
MDL Number: MFCD00148912
CAS Number: 50-99-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty