Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorosalicylic acid 98%

Supplier: AMBEED, INC
Warning

A238938-100G A238938-10G A238938-500G A238938-1000G
76976-394EA 20 USD InStock
76976-394 76976-396 76976-398 77672-140
4-Chlorosalicylic acid 98%
4-Chlorosalicylic acid
Precaución: for Research Use Only


Formula: C₇H₅ClO₃
MDL Number: MFCD00002449
CAS Number: 5106-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty