Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Spiro[2.5]octane-6-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A571534-1G A571534-5G A571534-100MG
76980-336EA 99 USD InStock
76980-336 76980-338 77673-534
Spiro[2.5]octane-6-carboxylic acid 97%
Spiro[2.5]octane-6-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₁₄O₂
MDL Number: MFCD11501504
CAS Number: 1086399-13-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty