Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 6,8-dibromoimidazo[1,2-a]pyrazine-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A185720-100MG A185720-1G A185720-250MG A185720-5G
76974-610EA 28 USD InStock
76974-610 76974-612 76974-614 76974-616
Ethyl 6,8-dibromoimidazo[1,2-a]pyrazine-2-carboxylate 95%
Ethyl 6,8-dibromoimidazo[1,2-a]pyrazine-2-carboxylate
Precaución: for Research Use Only


Formula: C₉H₇Br₂N₃O₂
MDL Number: MFCD14706101
CAS Number: 87597-21-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty