Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GSK461364 98+%

Supplier: AMBEED, INC
Warning

A313486-10MG A313486-1MG A313486-2MG A313486-5MG
76977-938EA 66 USD InStock
76977-938 76977-940 76977-942 76977-944
GSK461364 98+%
GSK461364
Precaución: for Research Use Only


Formula: C₂₇H₂₈F₃N₅O₂S
MDL Number: MFCD12755419
CAS Number: 929095-18-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty