Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Diethyl 2-aminosuccinate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A275053-10G A275053-1G A275053-25G
76977-262EA 23 USD InStock
76977-262 76977-264 76977-266
(R)-Diethyl 2-aminosuccinate hydrochloride 95%
(R)-Diethyl 2-aminosuccinate hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₆ClNO₄
MW: 225.66 g/mol
MDL Number: MFCD11225843
CAS Number: 112018-26-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty