Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-dimethylbut-3-enoic acid 95%

Supplier: AMBEED, INC
Danger

A296820-10G A296820-1G A296820-250MG A296820-5G
76977-666EA 337 USD InStock
76977-666 76977-668 76977-670 76977-672
2,2-dimethylbut-3-enoic acid 95%
2,2-dimethylbut-3-enoic acid
Precaución: for Research Use Only


Formula: C₆H₁₀O₂
MW: 114.144 g/mol
MDL Number: MFCD00196967
CAS Number: 10276-09-2
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty