Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-1H-pyrazol-5-amine 95%

Supplier: AMBEED, INC
Warning

A143105-100G A143105-25G A143105-500G
76972-688EA 123 USD InStock
76972-688 76972-690 76972-692
3-Methyl-1H-pyrazol-5-amine 95%
3-Methyl-1H-pyrazol-5-amine
Precaución: for Research Use Only


Formula: C₄H₇N₃
MW: 97.12 g/mol
MDL Number: MFCD00051640
CAS Number: 113402-89-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty