Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium 2,2-dimethyl-1,3-dioxolane-4-carboxylate 95%

Supplier: AMBEED, INC
Warning

A371671-100MG A371671-250MG A371671-1G
76978-712EA 20 USD InStock
76978-712 76978-714 77672-862
Potassium 2,2-dimethyl-1,3-dioxolane-4-carboxylate 95%
Potassium 2,2-dimethyl-1,3-dioxolane-4-carboxylate
Precaución: for Research Use Only


Formula: C₆H₉KO₄
MDL Number: MFCD08274678
CAS Number: 83400-91-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty