Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-(4-aminophenyl)propanoate 96%

Supplier: AMBEED, INC
Warning

A123951-100MG A123951-1G A123951-250MG
76971-722EA 20 USD InStock
76971-722 77571-444 77670-758
Methyl 3-(4-aminophenyl)propanoate 96%
Methyl 3-(4-aminophenyl)propanoate
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00546554
CAS Number: 35418-07-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty