Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Pyrazin-2-yl)acetonitrile 97%

Supplier: AMBEED, INC
Danger

A346548-100MG A346548-250MG
76978-478EA 49 USD InStock
76978-478 76978-480
2-(Pyrazin-2-yl)acetonitrile 97%
2-(Pyrazin-2-yl)acetonitrile
Precaución: for Research Use Only


Formula: C₆H₅N₃
MW: 119.12 g/mol
MDL Number: MFCD10697794
CAS Number: 5117-44-2
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty