Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Octan-2-yl 2-cyanoacetate 97%

Supplier: AMBEED, INC
Warning

A192125-100G A192125-10G A192125-1G A192125-25G A192125-500G A192125-5G
76974-882EA 55 USD InStock
76974-882 76974-884 76974-886 76974-888 76974-890 76974-892
Octan-2-yl 2-cyanoacetate 97%
Octan-2-yl 2-cyanoacetate
Precaución: for Research Use Only


Formula: C₁₁H₁₉NO₂
MDL Number: MFCD00236390
CAS Number: 52688-08-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty