Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-N-(3-([5,5'-Bipyrimidin]-2-ylamino)-4-methylphenyl)-4-((3-(dimethylamino)pyrrolidin-1-yl)methyl)-3-(trifluoromethyl)benzamide 98%

Supplier: AMBEED, INC
Warning

A156158-10MG A156158-1MG A156158-5MG
76973-322EA 66 USD InStock
76973-322 76973-324 76973-326
(S)-N-(3-([5,5'-Bipyrimidin]-2-ylamino)-4-methylphenyl)-4-((3-(dimethylamino)pyrrolidin-1-yl)methyl)-3-(trifluoromethyl)benzamide 98%
(S)-N-(3-([5,5'-Bipyrimidin]-2-ylamino)-4-methylphenyl)-4-((3-(dimethylamino)pyrrolidin-1-yl)methyl)-3-(trifluoromethyl)benzamide
Precaución: for Research Use Only


Formula: C₃₀H₃₁F₃N₈O
MW: 576.61 g/mol
MDL Number: MFCD11973647
CAS Number: 887650-05-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty