Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,9-Diphenyl-9H-carbazole-3-boronic acid 95%

Supplier: AMBEED, INC
Warning

A279064-1G A279064-250MG A279064-25G A279064-5G
76977-366EA 30 USD InStock
76977-366 76977-368 76977-370 76977-372
6,9-Diphenyl-9H-carbazole-3-boronic acid 95%
6,9-Diphenyl-9H-carbazole-3-boronic acid
Precaución: for Research Use Only


Formula: C₂₄H₁₈BNO₂
MDL Number: MFCD22123650
CAS Number: 1133058-06-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty