Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-2-ethyl-2H-indazole 96%

Supplier: AMBEED, INC
Warning

A665575-1G A665575-250MG A665575-5G
76980-884EA 37 USD InStock
76980-884 76980-886 76980-888
5-Amino-2-ethyl-2H-indazole 96%
5-Amino-2-ethyl-2H-indazole
Precaución: for Research Use Only


Formula: C₉H₁₁N₃
MDL Number: MFCD18375231
CAS Number: 5228-52-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty