Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-o-cresol 97%

Supplier: AMBEED, INC
Warning

A597141-100G A597141-1G A597141-25G A597141-5G
76980-476EA 212 USD InStock
76980-476 76980-478 76980-480 76980-482
5-Chloro-o-cresol 97%
5-Chloro-o-cresol
Precaución: for Research Use Only


Formula: C₇H₇ClO
MDL Number: MFCD00060672
CAS Number: 5306-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty