Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-fluoro-4-iodobenzoate 95%

Supplier: AMBEED, INC
Warning

A715862-100MG A715862-1G A715862-250MG
76981-206EA 8 USD InStock
76981-206 76981-208 76981-210
Ethyl-2-fluoro-4-iodobenzoate 95%
Ethyl 2-fluoro-4-iodobenzoate
Precaución: for Research Use Only


Formula: C₉H₈FIO₂
MW: 294.0615 g/mol
MDL Number: MFCD11846099
CAS Number: 205750-82-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty