Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-(trifluoromethyl)phenol 95%

Supplier: AMBEED, INC
Warning

A236533-10G A236533-1G A236533-25G A236533-5G
76976-358EA 14 USD InStock
76976-358 76976-360 76976-362 76976-364
3-Bromo-5-(trifluoromethyl)phenol 95%
3-Bromo-5-(trifluoromethyl)phenol
Precaución: for Research Use Only


Formula: C₇H₄BrF₃O
MDL Number: MFCD11041083
CAS Number: 1025718-84-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty