Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-8-bromooctanoate 99%

Supplier: AMBEED, INC
Warning

A351976-100G A351976-10G A351976-25G A351976-500G A351976-5G
76978-522EA 84 USD InStock
76978-522 76978-524 76978-526 76978-528 76978-530
Ethyl-8-bromooctanoate 99%
Ethyl-8-bromooctanoate
Precaución: for Research Use Only


Formula: C₁₀H₁₉BrO₂
MDL Number: MFCD00045057
CAS Number: 29823-21-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty