Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Eflornithine hydrochloride monohydrate 98%

Supplier: AMBEED, INC
Warning

A102810-100MG A102810-1MG A102810-50MG
76970-240EA 13 USD InStock
76970-240 76970-242 76970-244
Eflornithine hydrochloride monohydrate 98%
Eflornithine hydrochloride monohydrate
Precaución: For research use only.


Formula: C₆H₁₅ClF₂N₂O₃
MDL Number: MFCD00242734
CAS Number: 96020-91-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty