Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pemetrexed disodium heptahydrate 98%

Supplier: AMBEED, INC
Warning

A209789-1G A209789-25G
76975-494EA 25 USD InStock
76975-494 76975-496
Pemetrexed disodium heptahydrate 98%
Pemetrexed disodium heptahydrate
Precaución: for Research Use Only


Formula: C₂₀H₁₉N₅O₆·2Na·7H₂O
MW: 597.481 g/mol
MDL Number: MFCD09029394
CAS Number: 357166-29-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty