Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(N,N-Dimethylamino)phenylboronic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A113521-1G A113521-250MG A113521-5G
76971-020EA 7 USD InStock
76971-020 76971-022 76971-024
3-(N,N-Dimethylamino)phenylboronic acid hydrochloride 95%
3-(N,N-Dimethylamino)phenylboronic acid hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₃BClNO₂
MDL Number: MFCD04112542
CAS Number: 1256355-23-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty