Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4'-Dihydroxydiphenyl sulfone 95%

Supplier: AMBEED, INC
Warning

A234035-100G A234035-25G A234035-500G A234035-5G
76976-244EA 58 USD InStock
76976-244 76976-246 76976-248 76976-250
2,4'-Dihydroxydiphenyl sulfone 95%
2,4'-Dihydroxydiphenyl sulfone
Precaución: for Research Use Only


Formula: C₁₂H₁₀O₄S
MDL Number: MFCD01631305
CAS Number: 5397-34-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty