Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-2-(methylthio)pyrimidine-5-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A698618-100MG A698618-1G A698618-250MG A698618-5G
76981-066EA 12 USD InStock
76981-066 76981-068 76981-070 77673-908
4-Methoxy-2-(methylthio)pyrimidine-5-carboxylic acid 95%
4-Methoxy-2-(methylthio)pyrimidine-5-carboxylic acid
Precaución: for Research Use Only


Formula: C₇H₈N₂O₃S
MDL Number: MFCD02671593
CAS Number: 84332-06-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty