Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Chloroindole 98%

Supplier: AMBEED, INC
Warning

A715142-100MG A715142-1G A715142-250MG A715142-25G A715142-5G
76981-192EA 7 USD InStock
76981-192 76981-194 76981-196 76981-198 76981-200
7-Chloroindole 98%
7-Chloroindole
Precaución: for Research Use Only


Formula: C₈H₆ClN
MDL Number: MFCD01321301
CAS Number: 53924-05-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty