Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Bromo-5-chlorophenyl)ethanone 97%

Supplier: AMBEED, INC
Warning

A141012-10G A141012-5G A141012-1G
76972-618EA 41 USD InStock
76972-618 76972-620 77671-012
1-(2-Bromo-5-chlorophenyl)ethanone 97%
1-(2-Bromo-5-chlorophenyl)ethanone
Precaución: For research use only.


Formula: C₈H₆BrClO
MDL Number: MFCD11847057
CAS Number: 935-99-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty